Skip to Content
MilliporeSigma

SML3876

SHM115

≥98% (HPLC)

Synonym(s):

6-((Perfluorophenyl)amino)-[1,2,5]oxadiazolo[3,4-b]pyridin-5-ol, 6-[(2,3,4,5,6-Pentafluorophenyl)amino]-[1,2,5]oxadiazolo[3,4-b]pyridin-5(3H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H3F5N4O2
CAS Number:
Molecular Weight:
318.16
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=NC2=NON=C2C=C1NC3=C(C(F)=C(C(F)=C3F)F)F

Biochem/physiol Actions

Orally active mitochondria uncoupler that increases the energy expenditure and lowers the body fat mass in vivo.
SHM115 is an orally active mitochondria uncoupler (EC50 = 17 μM; measured as oxygen consumption rate (OCR) increase in L6 rat myoblast cells) that increases the energy expenditure and lowers the body fat mass in two diet-induced obesity mouse models, including an obesity prevention model (130 mg/kg/d via chow) and an obesity reversal model (130 mg/kg/d via oral gavage).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library