Skip to Content
MilliporeSigma

SML3877

SRI-32743

≥98% (HPLC)

Synonym(s):

2-(7-Methylimidazo[1,2-a]pyridin-6-yl)-N-(2-phenylbutyl)-4-quinazolinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H25N5
CAS Number:
Molecular Weight:
407.51
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Allosteric modulator of monoamine transporters that acts as partial antagonists of dopamine uptake.

SRI-32743 is an allosteric modulator of monoamine transporters that acts as partial antagonists of dopamine uptake. SRI-32743 attenuates Tat (the HIV regulatory protein transactivator of transcription) induced inhibition of DAT-mediated dopamine uptake. Also, it alleviates the potentiation of cocaine reward in HIV-1 Tat transgenic mice.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library