Skip to Content
MilliporeSigma

SML3889

NUCC-200823

≥98% (HPLC)

Synonym(s):

4,5,6,7-Tetrahydro-N-[[4-(1-piperidinyl)phenyl]methyl]-1-propyl-5-[[2-(4-pyridinyl)ethyl]amino]-1H-indazole-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H40N6O
CAS Number:
Molecular Weight:
500.68
NACRES:
NA.77
UNSPSC Code:
41116158
Assay:
≥98% (HPLC)
Form:
film or powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

film or powder

color

, Almost Colorless

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

NUCC-200823 is potent and selective agonists for ACKR3 that competed effectively with CXCL12 for binding to ACKR3. NUCC-200823 does not activate CXCR4 or D2 receptors
Potent and selective agonists for ACKR3 that competed effectively with CXCL12 for binding to ACKR3


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library