Skip to Content
MilliporeSigma

SML3892

CB-6644

≥98% (HPLC)

Synonym(s):

CB6644, 5-Chloro-2-ethoxy-4-fluoro-N-(4-((3-(methoxymethyl)-1-oxo-6,7-dihydro-1H,5H-benzo[c]pyrazolo[1,2-a][1,2]diazepin-2-yl)amino)-2,2-dimethyl-4-oxobutyl)benzamide, CB 6644

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H34ClFN4O5
CAS Number:
Molecular Weight:
573.06
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

CB-6644 is an orally active, non-ATP-competitive, potent and selective allosteric inhibitor against the RUVBL1/2 complex ATPase activity (IC50 = 15 nM; IC50 >100 μM against a panel of related AAA ATPases) with cancer killing potency in cultures (IC50 = 41-785 nM) and in xenograft myeloma models in vivo (150 mg/kg qd. or bid. via p.o.).
Orally active, potent and selective allosteric inhibitor against the RUVBL1/2 complex ATPase activity with cancer killing potency in cultures and in vivo.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library