Skip to Content
MilliporeSigma

Skip To

SML3913

VU0486846

≥98% (HPLC)

Synonym(s):

VU-0486846, (2R)-3,4-Dihydro-N-[(1S,2S)-2-hydroxycyclohexyl]-4-[[4-(1H-pyrazol-1-yl)phenyl]methyl]-2H-1,4-benzoxazine-2-carboxamide, (2R)-N-[(1S,2S)-2-Hydroxycyclohexyl]-4-[(4-pyrazol-1-ylphenyl)-methyl]-2,3-dihydro-1,4-benzoxazine-2-carboxamide, VU 0486846

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C25H28N4O3
CAS Number:
Molecular Weight:
432.51
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active, CNS-penetrant, muscarinic acetylcholine receptor M1-selective positive allosteric modulator (PAM) without agonist activity or adverse cholinergic toxicity.
VU0486846 is an orally active, CNS-penetrant and highly selective muscarinic acetylcholine receptor M1 positive allosteric modulator (PAM) with little agonist activity (rat M1 EC50 = 230 nM, 93% ACh max; agonist EC50 > 10 μM) or adverse cholinergic toxicity. VU0486846 improves pathology and cognitive deficits in female APPswe/PSEN1ΔE9 mice (10 mg/day via drinking water), as well as enhances recognition memory and reverses 3 mg/kg risperidone-induced deficits in acquisition of contextual fear conditioning in rats in vivo (3-10 mg/kg i.p.).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M9808
description

≥98% (HPLC)

description

affinity isolated antibody, lyophilized powder

assay

≥98% (HPLC)

assay

-

form

powder

form

lyophilized powder

Quality Level

100

Quality Level

200

storage temp.

-10 to -25°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

color

white to beige

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library