Skip to Content
MilliporeSigma

SML3917

(R)-SKBG-1

≥98% (HPLC)

Synonym(s):

(R)-4-(2-chloroacetyl)-N-(4-methoxybenzyl)-1-((4-methoxyphenyl)sulfonyl)piperazine-2-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H26ClN3O6S
CAS Number:
Molecular Weight:
495.98
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell permeable high affinity ligand of RNA-binding protein NONO; stabilize NONO–RNA interactions and rapidly reduces the expression of transcripts encoding AR and AR variants.


(R)-SKBG-1 is a cell permeable high affinity ligand of RNA-binding protein NONO that binds to Cys 145 of NONO and stabilize NONO–RNA interactions. (R)-SKBG-1 rapidly and stereoselectively reduces the expression of transcripts encoding the androgen receptor (AR) and its splice variants in prostate cancer cells.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library