Skip to Content
MilliporeSigma

SML3920

Q275

≥98% (HPLC)

Synonym(s):

2-[(2-Hydroxyphenyl)amino]-5-(2-methyl-3-phenyl-2-propen-1-ylidene)-4(5H)-thiazolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H16N2O2S
CAS Number:
Molecular Weight:
336.41
NACRES:
NA.77
UNSPSC Code:
51111800
eCl@ss:
32160703
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Potent, bacterial specific allosteric inhibitor of mitochondrial heme-copper oxidase (HCO).

Q275 is a potent, bacterial specific allosteric inhibitor of mitochondrial heme-copper oxidase (HCO). Q275 is an effective antibiotic against ceftriaxone-resistant Neisseria gonorrhoeae. Q275 does not affect the cellular respiration of mammalian cells.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library