Skip to Content
MilliporeSigma

SML3924

INCB3344

≥98% (HPLC)

Synonym(s):

INCB-3344, INCB 3344, trans-N-(2-{1-[4-(Benzo[d][1,3]dioxol-5-yl)-4-hydroxycyclohexyl]-4-ethoxypyrrolidin-3-ylamino}-2-oxoethyl)-3-(trifluoromethyl)benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H34F3N3O6
CAS Number:
Molecular Weight:
577.59
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active, potent and selective CCR2 antagonist in vitro and in vivo.
INCB3344 is an orally active, potent and selective CCR2 antagonist (human/mouse/rat/cynomolgus IC50 = 5.1/9.5/7.3/16 nM by competitive binding assays, IC50 = 3.8/7.8/2.7/6.2 nM by chemotaxis assays) with >100-fold selectivity over other homologous chemokine receptors. INCB3344 shows in vivo efficacy in a murine EAE model (thioglycolate-induced peritonitis in mice; 30-100 mg/kg bid. p.o. against peritoneal macrophage influx) and a rat adjuvant arthritis model (100 mg/kg bid. p.o. against adjuvant-induced macrophages infiltration of the joint).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library