Skip to Content
MilliporeSigma

SML3928

Bodilisant

≥98% (HPLC)

Synonym(s):

5,5-difluoro-1,3,7,9-tetramethyl-10-(4-(3-(piperidin-1-yl)propoxy)phenyl)-5H-4l4,5l4-dipyrrolo[1,2-c:2′,1′-f][1,3,2]diazaborinine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H34BF2N3O
CAS Number:
Molecular Weight:
465.39
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

red to red-black

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

High affinity histamine H3 receptor fluorescent ligand suitable to visualize H3 receptor in cells.



Bodilisant is a high affinity histamine H3 receptor fluorescent ligand suitable to visualize H3 receptor in cells. Bodilisant was successfully used to determine drug-target residence times for the H3 receptor ligands.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library