Skip to Content
MilliporeSigma

SML3935

icFSP1

≥95% (HPLC)

Synonym(s):

3,4,5-Trimethoxy-N-[4-(2-methyl-4-oxo-3(4H)-quinazolinyl)phenyl]benzeneacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H25N3O5
CAS Number:
Molecular Weight:
459.49
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=NC2=C(C(N1C3=CC=C(NC(CC4=CC(OC)=C(OC)C(OC)=C4)=O)C=C3)=O)C=CC=C2

Biochem/physiol Actions

Selective and efficacious in vivo inhibitor of ferroptosis-suppressor-protein 1 (FSP1); ferroptosis inducer.

icFSP1 is a selective and efficacious in vivo inhibitor of ferroptosis-suppressor-protein 1 (FSP1). icFSP1 triggers subcellular relocalization of FSP1 from the membrane and initiate FSP1 reversible aggregation into liquid droplet-like structures before ferroptosis initiation. It synergizes with ferroptosis inducing agents to potentiate the ferroptotic cell death response.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library