Skip to Content
MilliporeSigma

SML3943

LL-K9-3

≥98% (HPLC)

Synonym(s):

N-(5-(((5-(tert-Butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)-1-((2-(2-(((1R, 2S, 5R)-2-isopropyl-5-methylcyclohexyl)oxy)acetamido)ethyl)sulfonyl)piperidine-4-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H49N5O6S3
CAS Number:
Molecular Weight:
683.95
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=S(CCNC(CO[C@H]1[C@H](C(C)C)CC[C@@H](C)C1)=O)(N2CCC(C(NC3=NC=C(S3)SCC4=NC=C(O4)C(C)(C)C)=O)CC2)=O

InChI

1S/C31H49N5O6S3/c1-20(2)23-8-7-21(3)15-24(23)41-18-26(37)32-11-14-45(39,40)36-12-9-22(10-13-36)29(38)35-30-34-17-28(44-30)43-19-27-33-16-25(42-27)31(4,5)6/h16-17,20-24H,7-15,18-19H2,1-6H3,(H,32,37)(H,34,35,38)/t21-,23+,24-/m1/s1

InChI key

NPRIWHDFWDSJRZ-YFNKSVMNSA-N

Biochem/physiol Actions

Potent and selective CDK9-cyclin T1 complex degrader with stronger inhibition than THAL-SNS-032 against oncogenic transcription driven by Myc and AR.



LL-K9-3 is a potent and selective CDK9-cyclin T1 complex degrader (CDK9/CycT1 DC50 = 662/589 nM post 24h treatment, 22RV1 cells) with little potency toward other CDKs (1, 2, 4-7) or cyclins (E1, H, T2). LL-K9-3 exhibits higher antiproliferation activity than SNS-032 against 22RV1 prostate carcinoma cell (IC50 = 95 vs. 384 nM post 5-day treatment). Comparing to SNS-032 and THAL-SNS-032, LL-K9-3 shows stronger inhibition against oncogenic transcription driven by Myc and AR, as well as on several AR intrinsic target genes.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library