Skip to Content
MilliporeSigma

SML3956

Cdk5i SC-FT trifluoroacetate

≥95% (HPLC)

Synonym(s):

Cdk5i-FT negative control trifluoroacetate, Cdk5i-FT scrambled control trifluoroacetate, FITC-Ahx-AFRSPCARIGYV-TAT trifluoroacetate, FITC-Ahx-AFRSPCARIGYV-YGRKKRRQRRR (FITC = fluorescein isothiocyanate; Ahx = 6-aminohexanoate) trifluoroacetate, FITC-Ahx-Cdk5i SC-TAT trifluoroacetate, SC-FT trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C151H232N52O34S2
Molecular Weight:
3383.92
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

Biochem/physiol Actions

Scrambled control for the brain-penetrant and selective cyclin-dependent kinase Cdk5/p25 complex inhibitor Cdk5i-FT.

Cdk5i SC-FT is the scrambled control for Cdk5i-FT, a brain-penetrant and selective cyclin-dependent kinase Cdk5/p25 complex inhibitor in vitro and in vivo.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library