Skip to Content
MilliporeSigma

SML3958

AZ505

≥98% (HPLC)

Synonym(s):

AZ 505, AZ-505, N-Cyclohexyl-3-(3,4-dichlorophenethylamino)-N-(2-(2-(5-hydroxy-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-8-yl)ethylamino)ethyl)propanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H38Cl2N4O4
CAS Number:
Molecular Weight:
577.54
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, White to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C1NC(C(O)=CC=C2CCNCCN(C3CCCCC3)C(CCNCCC4=CC(Cl)=C(Cl)C=C4)=O)=C2OC1

InChI key

LIBVHXXKHSODII-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective SMYD2 inhibitor in vitro and in vivo.

AZ505 is a potent and selective SET and MYND domain-containing protein SMYD2 inhibitor (IC50 = 120 nM) that targets the substrate-binding site in a peptide substrate-competitive and cofactor SAM-uncompetitive manner, exhibiting no activity against lysine methyltransferases (KMTs) SMYD3, DOT1L, EZH2, GLP, G9A and SET7/9 (IC50 >83.3 µM). AZ505 downregulates p53 K370 monomethylation in U2OS cells (10h 10 µM) and delays cyst growth in early- (5 mg/kg/day i.p. from P7-P27) as well as later-stage (10 mg/kg via i.p. 3x per wk from P42-3.5 months, then 1x per wk from 3.5–6 months) Pkd1 conditional knockout mouse models in vivo.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library