Skip to Content
MilliporeSigma

SML3962

LTX-315 trifluoroacetate

≥95% (HPLC)

Synonym(s):

KKWWKKW(Dip)K-NH2 trifluoroacetate, LTX 315 trifluoroacetate, LTX-315 trifluoroacetate, Lys-Lys-Trp-Trp-Lys-Lys-Trp-Dip-Lys-NH2 (Dip = 3,3-diphenylalanine) trifluoroacetate, Ruxotemitide trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C78H106N18O9
Molecular Weight:
1439.79
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

[nH]1c2c(c(c1)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CCCCN)CCCCN)Cc7c8c([nH]c7)cccc8)Cc5c6c([nH]c5)cccc6)CCCCN)CCCCN)C(=O)N[C@@H](C(c4ccccc4)c3ccccc3)C(=O)N[C@@H](CCCCN)C(=O)N)cccc2

InChI key

GGAKLYWEFZCVIT-TVEKFXMRSA-N

Biochem/physiol Actions

Mitochondrial membrane-permeabilizing cationic oncolytic peptide with cancer-selective cytotoxicity in cultures and in vivo.


LTX-315 (Ruxotemitide) is a 9-mer cationic oncolytic peptide that preferentially permeabilizes mitochondrial membranes, thereby causing partially BAX/BAK1-regulated, caspase-independent necrosis. LTX-315 exhibits cancer-selective cytotoxicity (IC50 = 1.2-7.2 μM among 47 cancer lines; normal MRC-5/HUVEC-C/RBC IC50 = 9.2/16.3/695 μM) with low inhibitory potency toward CYP450 enzymes (IC50 from 7.9 to >100 μM). Intratumoral administration results in B16 melanoma tumor necrosis and the infiltration of immune cells into the tumor parenchyma followed by complete tumor regression in mice in vivo (1 mg/50 μL i.t.). Animals cured by LTX-315 treatment are protected against B16 tumor cells re-challenge.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library