Skip to Content
MilliporeSigma

SML3973

Fidarestat

≥98% (HPLC)

Synonym(s):

(2S,4S)- 6-Fluoro-2,3-dihydro-2′,5′-dioxo-spiro[4H-1-benzopyran-4,4′-imidazolidine]-2-carboxamide, (2S,4S)-6-Fluoro-2′,5′-dioxospiro[chroman-4,4′-imidazolidine]-2-carboxamide, SNK-860

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H10FN3O4
CAS Number:
Molecular Weight:
279.22
MDL number:
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Active site-targeting, potent and selective aldose reductase (AR) inhibitor with >100-fold selectivity over aldehyde reductase (AHR).



Fidarestat inhibits Aldose Reductase (AR) involved in the rate limiting step in the conversion of glucose to sorbitol under hyperglycemic conditions. Aldose reductase has been the target of multiple clinical investigatons to treat diabetic neuropathy and retinopathy. Fidarestat has recently been shown to prevent inflammatory signals (TNF-α, LPS) that cause cancer (colon, breast, prostate and lung), metastasis, asthma, and other inflammatory diseases, as well as being useful for treatment of diabetic neuropathy.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library