Skip to Content
MilliporeSigma

SML3980

HHS-465 10% solution in DMSO

≥98% (HPLC)

Synonym(s):

N-2-Propyn-1-yl-4-(2H-1,2,3-triazol-2-ylsulfonyl)benzamide 10% solution in DMSO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H10N4O3S
Molecular Weight:
290.30
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
(10% Solution in DMSO)
Storage condition:
(Material decomposes above 90°C, keep below 40°C during handling)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(10% Solution in DMSO)

storage condition

(Material decomposes above 90°C, keep below 40°C during handling)

color

white to beige

shipped in

wet ice

storage temp.

-10 to -25°C

SMILES string

O=S(N1N=CC=N1)(C2=CC=C(C(NCC#C)=O)C=C2)=O

Biochem/physiol Actions

Cell penetrant tyrosine/lysine-reactive (Y-and K-reactive) sulfonyl-triazole exchange (SuTEx) probe.
HHS-465 is a cell penetrant tyrosine/lysine-reactive (Y-and K-reactive) sulfonyl-triazole exchange (SuTEx) probe suitable to study novel nucleophilic sites on established and poorly described protein targets.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library