Skip to Content
MilliporeSigma

SML3996

Pentachloropseudilin

≥98% (HPLC)

Synonym(s):

2,4-Dichloro-6-(3,4,5-trichloro-1H-pyrrol-2-yl)phenol, 2,4-Dichloro-6-(3,4,5-trichloro-2-pyrrolyl)phenol, Antibiotic A 15104Y, PClP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H4Cl5NO
CAS Number:
Molecular Weight:
331.41
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

OC1=C(C2=C(Cl)C(Cl)=C(Cl)N2)C=C(Cl)C=C1Cl

InChI

1S/C10H4Cl5NO/c11-3-1-4(9(17)5(12)2-3)8-6(13)7(14)10(15)16-8/h1-2,16-17H

InChI key

FBRLLYYPGGXCKT-UHFFFAOYSA-N

Biochem/physiol Actions

Reversible allosteric inhibitor of myosin ATPase and motor activity specific for class-1 myosin; inhibits T. cruzi parasite motility.
Pentachloropseudilin is a reversible allosteric inhibitor of myosin ATPase and motor activity specific for class-1 myosin. Pentachloropseudilin is a natural product present in marine bacteria Chromobacterium sp., Pseudomonas bromoutilis, and Alteromonas luteoviolaceus. Pentachloropseudilin inhibits T. cruzi parasite motility, host cell adhesion and invasion.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library