Skip to Content
MilliporeSigma

SML4003

TP-060

≥98% (HPLC)

Synonym(s):

2-(4-Fluoro-2-(2,2,2-trifluoroethoxy)phenyl)-8-(4-(1-hydroxy-1-methylethyl)phenyl)-3,8-diaza-bicyclo[4.3.0]nona-1(6),2,4-trien-7-one, 4-[4-Fluoro-2-(2,2,2-trifluoroethoxy)phenyl]-2,3-dihydro-2-[4-(1-hydroxy-1-methylethyl)phenyl]-1H-pyrrolo[3,4-c]pyridin-1-one, 4-[4-Fluoro-2-(2,2,2-trifluoroethoxy)phenyl]-2-[4-(2-hydroxypropan-2-yl)phenyl]-2,3-dihydro-1H-pyrrolo[3,4-c]pyridin-1-one, Glucosylceramide synthase-IN-1, T 036, T-036, T036, TP 060, TP060

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H20F4N2O3
CAS Number:
Molecular Weight:
460.42
UNSPSC Code:
12352202
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C1C2=C(CN1C3=CC=C(C(C)(C)O)C=C3)C(C4=C(OCC(F)(F)F)C=C(F)C=C4)=NC=C2

InChI key

OAZAPIKYVGRNCO-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, brain-penetrant, substrate-noncompetitive glucosylceramide synthase (GCS) inhibitor with therapeutic efficacy in a murine Gaucher’s disease (GD) model in vivo.
TP-060 (T-036) is an orally active, brain-penetrant, substrate-noncompetitive, potent and selective glucosylceramide synthase (GCS) inhibitor (human/mouse IC50 = 31/51 nM) that effectively downregulates glucosylceramide (GlcCer) level in GBA mutant human fibroblasts containing mutated glucosylceramidase (IC50 = 7.6 nM). TP-060 is efficacious in lowering both GlcCer and glucosylsphingosine (GlcSph) levels in the plasma and cerebral cortex of Gba D409V KI mice in vivo following a two-month treatment in the murine Gaucher’s disease (GD) model (10 and 30 mg/kg/day p.o.).


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library