Skip to Content
MilliporeSigma

SML4016

PF-9363

≥98% (HPLC)

Synonym(s):

2,6-Dimethoxy-N-[4-methoxy-6-(1H-pyrazol-1-ylmethyl)-1,2-benzisoxazol-3-yl]benzenesulfonamide, CTx 648, CTx-648, CTx648, PF 9363, PF9363

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H20N4O6S
CAS Number:
Molecular Weight:
444.46
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=S(NC1=NOC2=CC(CN3N=CC=C3)=CC(OC)=C21)(C4=C(OC)C=CC=C4OC)=O

InChI

1S/C20H20N4O6S/c1-27-14-6-4-7-15(28-2)19(14)31(25,26)23-20-18-16(29-3)10-13(11-17(18)30-22-20)12-24-9-5-8-21-24/h4-11H,12H2,1-3H3,(H,22,23)

InChI key

VNSQFPFWUAZOTQ-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, highly potent and selective lysine acetyltransferase KAT6A inhibitor with anti-cancer efficacy in vivo.



PF-9363 (CTx-648) is an orally active, highly potent and selective lysine acetyltransferase KAT6A inhibitor (KAT6A Ki = 0.27 nM; KAT6B/7/5/8 Ki = 2.4/70/420/670 nM). PF-9363 selectively inhibits H3K23 acetylation (ZR-75-1 H3K23Ac/H3K14Ac IC50 = 0.85/120 nM) and exhibits anti-proliferation potency against high KAT6A-expressing estrogen receptor signaling-positive (ER+) breast cancer cells in cultures (ZR-75-1 IC50 = 0.37 nM) and in mice in vivo (complete inhibition of ZR-75-1 xenograft tumor growth by 0.03 mg/kg/day p.o.) by downregulating cancr ER signaling, cell cycle and MYC transcriptional programs.


Still not finding the right product?

Explore all of our products under PF-9363


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library