Skip to Content
MilliporeSigma

SML4018

VER-246608

≥98%

Synonym(s):

N-[4-(2-Chloro-5-methyl-4-pyrimidinyl)phenyl]-N-[[4-[[(2,2-difluoroacetyl)amino]methyl]phenyl]methyl]-2,4-dihydroxybenzamide, N-[4-(2-Chloro-5-methylpyrimidin-4-yl)phenyl]-N-([4-[(2,2-difluoroacetamido)methyl]phenyl]methyl)-2,4-dihydroxybenzamide, VER 246608

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H23ClF2N4O4
CAS Number:
Molecular Weight:
552.96
MDL number:
NACRES:
NA.21
Assay:
≥98%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C(N(CC1=CC=C(CNC(C(F)F)=O)C=C1)C2=CC=C(C3=NC(Cl)=NC=C3C)C=C2)C4=CC=C(C=C4O)O

Biochem/physiol Actions

An ATP-competitive, potent pan-PDK inhibitor that efficiently disrupts Warburg metabolism
VER-246608 is a cell-permeable resorcinol amide compound that acts as a potent, selective, reversible, and ATP-competitive inhibitor of pan-PDK isoform PDK (IC50 = 35, 84, 40 and 91 nM against PDK1, PDK2, PDK3 and PDK4, respectively) with minimal activity towards HSP-90 (IC50 > 10 µM). VER-246608 effectively enhances PDHA1 (pyruvate dehydrogenase E1-alpha subunit) activity and markedly diminishes PDHA1-Ser293 and -Ser300 phosphorylations without inducing severe cancer cell death. Efficiently disrupts Warburg metabolism and demonstrates cancer cells growth arrest under nutrient-deficient conditions. Also inhibits tumor spheroids growth with a potency that was comparable to cells grown in 2D-culture.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library