Skip to Content

SML4027

ZH8659 hydrochloride

≥98% (HPLC)

Synonym(s):

2-[4-(Furan-2-yl)phenyl]ethan-1-amine hydrochloride, 4-(2-Furanyl)benzene-ethanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H13NO ·HCl
Molecular Weight:
223.70
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NCCC1=CC=C(C2=CC=CO2)C=C1.Cl

InChI

1S/C12H13NO.ClH/c13-8-7-10-3-5-11(6-4-10)12-2-1-9-14-12;/h1-6,9H,7-8,13H2;1H

InChI key

PIDHDDFXZYDGMH-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available, potent and selective agonist of Trace amine-associated receptor 1 (TAAR1); potent Gq-selective agonist of mTAAR1/hTAAR1.

ZH8659 is an orally available, potent and selective agonist of Trace amine-associated receptor 1 (TAAR1). ZH8659 is a potent Gq-selective agonist of mTAAR1/hTAAR1 (EC50 = 2.6 nM (Gq)). ZH8659 alleviates MK-801-induced schizophrenia-like phenotypes without inducing catalepsy in mice.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library