Skip to Content
MilliporeSigma

SML4034

BODIPY 630/650 X NHS ester

≥85% (HPLC)

Synonym(s):

(T-4)-[2,5-Dioxo-1-pyrrolidinyl 6-[[2-[4-[2-[5-[[5-(2-thienyl)-2H-pyrrol-2-ylidene-κN]methyl]-1H-pyrrol-2-yl-κN]ethenyl]phenoxy]acetyl]amino]hexanoato]difluoroboron, BODIPY 630/650X, BODIPY 630/650 NHS Ester, Bodipy 630/650 succinimidyl ester, BODIPY-X 630/650-NHS ester, BODIPY 630/650X SE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H31BF2N4O6S
CAS Number:
Molecular Weight:
660.50
MDL number:
NACRES:
NA.21
Assay:
≥85% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥85% (HPLC)

form

powder

color

purple to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(NCCCCCC(ON1C(CCC1=O)=O)=O)COC(C=C2)=CC=C2/C=C/C3=CC=C4N3[B-](F)(F)[N+]5=C(C6=CC=CS6)C=CC5=C4

Biochem/physiol Actions

Amine reactive, red fluorescent labeling reagent that can be used to tag antibodies, proteins, peptides, lipids, and nucleic acids.

BODIPY-X 630/650-NHS ester, a derivative of Boron-dipyrromethene (BODIPY), features a seven-atom aminohexanoyl spacer, is a versatile, amine-sensitive red fluorescent labeling reagent useful to tag antibodies, proteins, peptides, lipids, and nucleic acids. BODIPY 630/650 X is characterized by low toxicity and can be excited at approximately 630 nm, emitting fluorescence at around 650 nm. BODIPY 630/650X stands out due to its high molar extinction coefficient, impressive quantum yield, and an extended lifetime of its excited state. Furthermore, BODIPY-X 630/650 is notable for its brightness, remarkable photostability, and distinct emission spectrum. These properties minimize spectral overlap with other fluorophores and enhance its utility in multiplex imaging, where multiple fluorophores are used simultaneously.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library