Skip to Content
MilliporeSigma

SML4037

ASP7967

≥98% (HPLC)

Synonym(s):

N2,N4-bis(4-Fluorophenyl)-N6-(2-pyrimidinylmethyl)-1,3,5-triazine-2,4,6-triamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H16F2N8
CAS Number:
Molecular Weight:
406.39
UNSPSC Code:
41106305
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(NC2=NC(NCC3=NC=CC=N3)=NC(NC4=CC=C(C=C4)F)=N2)C=C1

InChI key

UAAKDZMQDFXQTN-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable, low-toxic triazine-triamine ligand of the RNA aptamer AC17-4 that effectively activates gene expression in riboswitches and significantly raises hEPO levels in mice.

ASP7967 is a cell permeable, low-toxic triazine-triamine ligand of the RNA aptamer AC17-4 with a Kd of 12 nM and an EC50 of 2.3 µM. ASP-7967 significantly activates gene expression in aptazyme-based riboswitches, exceeding a tenfold increase in HEK293 cells. Furthermore, when administered orally at 100 mg/kg, ASP 7967 substantially boosts serum human erythropoietin (hEPO) levels in mice, about 7.2 times more than the control, demonstrating its robust in vivo effectiveness.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library