Skip to Content
MilliporeSigma

SML4041

CNTinh-03

≥98% (HPLC)

Synonym(s):

2-[[(3-carboxybicyclo[2.2.1]hept-2-yl)carbonyl]amino]-5-methyl-4-(4-methylphenyl)-3-thiophenecarboxylic acid 3-methyl ester, 3-methoxycarbonyl-5-methyl-4-p-toyly-2-aminothiophene-bicyclo[2.2.1]heptane-2-carboxylic acid, FM183377

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H25NO5S
CAS Number:
Molecular Weight:
427.51
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Cyclic nucleotide suppressor.
CNTinh-03 is a small-molecule general suppressors of cellular cyclic nucleotides. It reduces basal and agonist-induced elevation of cytoplasmic cAMP and cGMP in multiple cells and in response to different types of agonists. The mechanism of action involves nonselective activation of phosphodiesterase (PDE).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library