Skip to Content
MilliporeSigma

SML4043

PCI-24781

≥98% (HPLC)

Synonym(s):

3-[(Dimethylamino)methyl]-N-[2-[4-(hydroxycarbamoyl)phenoxy]ethyl]-1-benzofuran-2-carboxamide, 3-[(Dimethylamino)methyl]-N-[2-[4-[(hydroxyamino)carbonyl]phenoxy]ethyl]-2-benzofurancarboxamide, Abexinostat, CRA-024781

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H23N3O5
CAS Number:
Molecular Weight:
397.42
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N(O)C(=O)c1ccc(cc1)OCCNC(=O)c2[o]c3c(c2CN(C)C)cccc3, CN(C)CC1=C(C(NCCOC2=CC=C(C(NO)=O)C=C2)=O)OC3=CC=CC=C31

InChI

1S/C21H23N3O5/c1-24(2)13-17-16-5-3-4-6-18(16)29-19(17)21(26)22-11-12-28-15-9-7-14(8-10-15)20(25)23-27/h3-10,27H,11-13H2,1-2H3,(H,22,26)(H,23,25)

InChI key

MAUCONCHVWBMHK-UHFFFAOYSA-N

Biochem/physiol Actions

Orally bioavailable, potent and selective pan-HDAC inhibitor.
PCI-24781 (Abexinostat) is an orally bioavailable, potent, and selective pan-HDAC inhibitor that exhibits potent antitumor activities. It inhibits all Class I and Class II HDAC isoforms (Ki = 7 nM HDAC1, 19 nM HDAC2, 8.2 nM HDAC3/SMRT, 17 nM HDAC6, 280 nM HDAC8, 24 nM HDAC10) although it is reported to be a more potent inhibitor of HDACs 1 and 3 at low concentrations.


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library