Skip to Content
MilliporeSigma

SML4044

TRC051384

≥98% (HPLC)

Synonym(s):

1-[2-(Morpholin-4-yl)ethyl]-3-(4-{-3-[6-(morpholin-4-yl)pyridin-2-yl]prop-2-enoyl}phenyl)urea, N-[2-(4-Morpholinyl)ethyl]-N′-[4-[3-[6-(4-morpholinyl)-2-pyridinyl]-1-oxo-2-propen-1-yl]phenyl]urea, TRC 051384 free base, TRC-051384 free base

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H31N5O4
CAS Number:
Molecular Weight:
465.54
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(NC(NCCN2CCOCC2)=O)C=C1)/C=C/C3=CC=CC(N4CCOCC4)=N3

Biochem/physiol Actions

Potent heat shock protein 70 inducer with therapeutic efficacy in a rat model of transient cerebral ischemia in vivo.
TRC051384 is a potent heat shock protein 70 inducer (fold of HSP70 mRNA induction in 4h at 6.25/12.5 µM = 19/210 in HeLa & 10/84 in primary neonatal rat cerebellum neurons) that increases HSF1 transcriptional activity in HeLa cells (by 2.1/3.3-fold at 12.5/2.5 µM) and reduces LPS-induced TNF-α expression in THP-1 cells (% inhibition/conc. = 60/6.25 µM & 90/12.5 µM). TRC051384 reduces neuronal injury and disability in a rat model of transient cerebral ischemia in vivo (9 mg/kg 4h or 8h post onset of ischemia, then 4.5 mg/kg every 2h for 48h via i.p.).


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library