Skip to Content
MilliporeSigma

SML4049

TT-012, MITF dimer disruptor

≥98% (HPLC)

Synonym(s):

(2E,2′E)-N,N′-(Pyridine-2,6-diyl)bis(3-(furan-2-yl)acrylamide), (2E,2′E)-N,N′-2,6-Pyridinediylbis[3-(2-furanyl)-2-propenamide], TT 012, TT012

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H15N3O4
CAS Number:
Molecular Weight:
349.34
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC(NC(/C=C/C2=CC=CO2)=O)=N1)/C=C/C3=CC=CO3

InChI

1S/C19H15N3O4/c23-18(10-8-14-4-2-12-25-14)21-16-6-1-7-17(20-16)22-19(24)11-9-15-5-3-13-26-15/h1-13H,(H2,20,21,22,23,24)/b10-8+,11-9+

InChI key

YUFJMFXVUUMVCA-GFULKKFKSA-N

Biochem/physiol Actions

Potent and selective MITF dimerization disruptor with significant efficacy against melanoma both in vitro and in vivo.
TT-012 is a highly effective microphthalmia-associated transcription factor (MITF) dimerization inhibitor (Kd = 15.5 nM for the bHLH-LZ domain) that specifically targets melanoma by disrupting MITF′s DNA-binding activity. TT012 exhibits notable potency in inhibiting MITF dimer formation (IC50 = 13.1 nM) and reducing the growth of B16F10 melanoma cells (IC50 = 499 nM), while also lowering mRNA levels of MITF target genes in these cells (IC50 < 3.12 µM). TT 012 is particularly effective against melanoma cells with elevated MITF expression and markedly diminishes tumor growth and pulmonary metastasis in mice, achieving reductions of 79.7% and 93.9% at dosages of 2 mg/kg and 5 mg/kg, respectively, compared to controls.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library