Skip to Content
MilliporeSigma

SML4050

OTUB2-IN-1

≥98% (HPLC)

Synonym(s):

(E)-6-(5-(1-(Carboxymethyl)-2-oxoindolin-3-ylidene)-4-oxo-2-thioxothiazolidin-3-yl)hexanoic acid, OTUB2 inhibitor, OTUB2i

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18N2O6S2
CAS Number:
Molecular Weight:
434.49
UNSPSC Code:
51111800
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, Light orange to very dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N(C1=C/2C=CC=C1)CC(O)=O)C2=C3C(N(C(S/3)=S)CCCCCC(O)=O)=O

InChI key

VQTUGMPRAUNMNP-FOCLMDBBSA-N

Biochem/physiol Actions

Inhibits tumor growth by selectively disrupting OTUB2′s activity, thereby reducing PD-L1 levels and enhancing antitumor immunity.
OTUB2-IN-1 selectively targets OTUB2 (OTU deubiquitinase, ubiquitin aldehyde binding 2), a positive regulator of PD-L1, to diminish PD-L1 levels dose-dependently in tumor cells, thereby enhancing antitumor immunity. OTUB2i binds reversibly to OTUB2′s catalytic pocket (KD = 12 µM), selectively impairing its deubiquitinase function without disrupting OTUB2-PD-L1 interactions. OTUB2-IN-1 operates by destabilizing PD-L1 through Lys-48-linked polydeubiquitination leading to a significant reduction in PD-L1-overexpressing tumor growth and enhanced CD8+ CTL-mediated tumor cell elimination, increasing T-cell infiltration in tumors, all achieved without noticeable toxicity. Note: The recommended usage of OTUB2-IN-1 is 10 µM for cellular applications, and for in vivo experiments, it is suggested at 20 mg/kg, administered intraperitoneally (i.p.) in mice. This dosing strategy aims to maximize the inhibitor′s effectiveness in reducing PD-L1 levels in tumor cells while minimizing potential toxicity.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library