Skip to Content
MilliporeSigma

SML4052

GSK023

≥98% (HPLC)

Synonym(s):

(S)-5-(1-((1-(1-Isopropylpiperidine-4-carbonyl)piperidin-3-yl)-methyl)-1H-benzo[d]imidazole-2-yl)-1,3-dimethylpyridin-2(1H)-one, GSK 023, GSK-023

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H39N5O2
Molecular Weight:
489.65
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

GSK023 is a potent and selective probe for the N-terminal bromodomain (BD) BD1 of bromodomain and extra-terminal (BET) proteins (pIC50 & BD1/BD2 fold selectivity = 8.0/400/BRD2, 7.7/400/BRD3, 7.8/1000/BRD4, 7.3/630/BRDT by TR-FRET; pKd & BD1/BD2 fold selectivity = 8.5/1000/BRD2, 8.4/400/BRD3, 8.1/316/BRD4, 8.1/400/BRDT by BROMOscan; pKd = 5.0/EP300, >2000/20 non-BET BDs) with antiproliferative potency against human primary T cells, coronary artery smooth muscle cells, endothelial cells, and fibroblasts (0.16-10 µM) by impacting a variety of immune-modulatory biomarkers (MCP-1 pIC50 = 6.9 using human whole blood).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library