Skip to Content
MilliporeSigma

SML4060

NIAD-4

≥98% (HPLC)

Synonym(s):

2-((5′-(4-Hydroxyphenyl)-2,2′-bithiophen-5-yl)methylene)malononitrile, 2-[[5′-(4-Hydroxyphenyl)[2,2′-bithiophen]-5-yl]methylene]propanedinitrile, 2-[[5-[5-(4-Hydroxyphenyl)thiophen-2-yl]thiophen-2-yl]methylidene]propanedinitrile, 5’-(4-Hydroxyphenyl)-5-(2,2-dicyanoethenyl)-2,2’-bithiophene, NIAD 4, NIAD4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H10N2OS2
CAS Number:
Molecular Weight:
334.41
MDL number:
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, Red to very dark red-brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(S3)C=C(C#N)C#N

Biochem/physiol Actions

Blood-brain barrier (BBB)-permeable, far-red (near-IR) fluorescent contrast agent for staining of amyloid plaques with high specificity in situ and in vivo.

NIAD-4 is a blood-brain barrier (BBB)-permeable, far-red (near-IR) fluorescent contrast agent for staining of amyloid plaques with high specificity in situ (10 μM in DMSO/propylene glycol; APP mice coronal section) and in vivo (2 mg/kg APP mice i.v.). Due to its push-pull structure design and a greatly enhanced thioflavin T (ThT) site affinity (Ki = 10 nM/NIAD-4 vs 580 nM/ThT), NIAD-4 exhibits significantly red-shifted (~70 nm) absorption and a 1.3-fold increase in extinction coefficient upon binding aggregated amyloid-beta 1-40 (4.1 μM NIAD-4 & 10 μM amyloid-β in PBS). Ex 450-488 nm, Em 550-650 nm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library