Skip to Content
MilliporeSigma

SML4067

MCLA Hydrochloride

≥98% (HPLC)

Synonym(s):

2-Methyl-6-(p-methoxyphenyl)-3,7-dihydroimidazo[1,2-a]pyrazin-3-one HCl, 3,7-Dihydro-2-methyl-6-(4-methoxyphenyl)imidazol[1,2-a]pyrazine-3-one HCl, 6-(4-Methoxyphenyl)-2-methyl-3,7-dihydroimidazo[1,2-a]pyrazin-3-one HCl, 6-(4-Methoxyphenyl)-2-methylimidazo[1,2-a]pyrazin-3(7H)-one hydrochloride, MCLA HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H14ClN3O2
CAS Number:
Molecular Weight:
291.73
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, Faint yellow to very dark orange

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C1C(C)=NC2=CNC(C3=CC=C(OC)C=C3)=CN21.Cl

Biochem/physiol Actions

Chemiluminescence probe, after glow substrate, known for its high sensitivity in detecting reactive oxygen species like superoxide anion and singlet oxygen, both intracellularly and extracellularly.

MCLA (methoxylated Cypridina luciferin analog) is a highly sensitive probe employed in the detection of reactive oxygen species (ROS), particularly superoxide anion (O2•−) and singlet oxygen (1O2). It is notable for its blue luminescence (~ 455 nm), which is a critical indicator in ROS detection. MCLA′s detection mechanism involves oxidation to a probe radical, which then reacts with O2•− to produce luminescence. MCLA offers a broad dynamic range, making it versatile for both intracellular measurements in live cells and extracellular detection in various sample types. Its application is particularly significant in research related to oxidative stress, inflammation, and neurodegenerative diseases, where ROS play a pivotal role. Ex: ~ 430 nm, Em: ~ 549 nm.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library