Skip to Content
MilliporeSigma

SML4071

BAY-593

≥95% (HPLC)

Synonym(s):

(R)-3,3,3-Trifluoro-2-methoxy-1-((R)-7-(2-methoxyphenyl)-3,9-diazaspiro[5.5]undecan-3-yl)-2-phenylpropan-1-one, (2R)-3,3,3-Trifluoro-2-methoxy-1-[(7R)-7-(2-methoxyphenyl)-3,9-diazaspiro[5.5]undec-3-yl]-2-phenyl-1-propanone, BAY 593, BAY593, GGTase-I inhibitor BAY-593, YAP1/TAZ pathway inhibitor BAY-593

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H31F3N2O3
CAS Number:
Molecular Weight:
476.53
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

BAY-593, a GGTase-I inhibitor, primarily targets the YAP1/TAZ signaling pathway, a crucial axis in cell proliferation and tumor progression. Notably, BAY593 alters the mRNA expression levels of YAP1/TAZ target genes including CYR61, CTCF, ANKRD1, and CD274, indicating its specific molecular action. In vitro studies reveal its IC50 values: 9.41 nM for MDA-MB-231 TEAD-luc, 0.564 µM for MDA-MB-231 proliferation, and 38.4 nM for HT-1080 proliferation. The efficacious in vivo dose was determined as 5 mg/kg (p.o., BID), in the PXF 541 pleuromesothelioma xenograft model, showcasing its therapeutic potential in cancer treatment.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library