Skip to Content
MilliporeSigma

SML4083

TJ-M2010-5

≥98% (HPLC)

Synonym(s):

3-(4-Benzyl-piperazin-1-yl)-N-(4-phenylthiazol-2-yl)propionamide, 3-(4-Benzylpiperazin-1-yl)-N-(4-phenylthiazol-2-yl)propanamide, 4-(Phenylmethyl)-N-(4-phenyl-2-thiazolyl)-1-piperazinepropanamide, TJ-5, TJM2010-5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H26N4OS
CAS Number:
Molecular Weight:
406.54
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCN1CCN(CC1)CC2=CC=CC=C2)NC3=NC(C4=CC=CC=C4)=CS3

Biochem/physiol Actions

TIR domain-targeting MyD88 homodimerization inhibitor that blocks MyD88 signaling and prevents colitis-associated cancer (CAC) development in vivo.

TJ-M2010-5 (TJ-5) is a TIR domain-targeting MyD88 homodimerization inhibitor (67% inhibition of HEK293 cellular MyD88 interaction at 20 µM) that blocks IRAK4, ERK and p38 phosphorylation induction by LPS stimulation (100 ng/mL for 6h with or without 40 µM TJ-5) in RAW 264.7 cells. TJ-5 (50 mg/kg/d i.p.) significantly reduces AOM/DSS-induced colitis and completely prevents colitis-associated cancer (CAC) development and death (53% mortality of control mice) in mice in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library