Skip to Content
MilliporeSigma

SML4084

N6-Furfuryladenosine

≥98% (HPLC)

Synonym(s):

Kinetin riboside, Ribosylkinetin, NSC 120958, N-(2-Furanylmethyl)-adenosine, 6-Furfurylaminopurine riboside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H17N5O5
CAS Number:
Molecular Weight:
347.33
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352207
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@@]1([C@@]([H])([C@]([H])(O[C@@]1([H])N2C=NC3=C2N=CN=C3NCC4=CC=CO4)CO)O)[H]

Biochem/physiol Actions

Naturally occurring purine nucleoside analog that protect cells against oxidative stress.
N6-Furfuryladenosine (Kinetin riboside) is a purine nucleoside analog. It is a naturally occurring plant cytokine and a product of oxidative DNA damage. N6-Furfuryladenosine protect cells against oxidative stress and delay of age-related phenotypes in human fibroblasts. N6-furfuryladenosine enhances mitochondrial turnover and bioenergetics in the diabetic mouse retina (Ins2Akita males). N6-Furfuryladenosine displays strong antiproliferative activity in various human cancer cells.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library