Skip to Content
MilliporeSigma

SML4087

Spns2 inhibitor 16d

≥98% (HPLC)

Synonym(s):

3-(4-Decylphenyl)-1,2,4-oxadiazole-5-propanamine hydrochloride, SLF1081851

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H33N3O·HCl
CAS Number:
Molecular Weight:
379.97
UNSPSC Code:
41106300
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC1=CC=C(C=C1)C2=NOC(CCCN)=N2.Cl

InChI

1S/C21H33N3O.ClH/c1-2-3-4-5-6-7-8-9-11-18-13-15-19(16-14-18)21-23-20(25-24-21)12-10-17-22;/h13-16H,2-12,17,22H2,1H3;1H

InChI key

ZJJXKBLUUGLWDW-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective inhibitor of Spns2 (spinster homolog 2) sphingosine-1-phosphate (SP1) transporter.
Spns2 inhibitor 16d (SLF1081851) is a potent and selective inhibitor of Spns2 (spinster homolog 2) sphingosine-1-phosphate (SP1) transporter. Spns2 inhibitor 16d (SLF1081851) decreases circulating plasma S1P and lymphocytes in mice and rats. Apparently, it locks Spns2 in the inward-facing state.

Disclaimer

Hygroscopic


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library