Skip to Content
MilliporeSigma

SML4095

Motolimod

≥98% (HPLC)

Synonym(s):

VTX 378, VTX-2337, 2-Amino-N,N-dipropyl-8-[4-(1-pyrrolidinylcarbonyl)phenyl]-3H-1-benzazepine-4-carboxamide, 2-Amino-N,N-dipropyl-8-[4-(pyrrolidine-1-carbonyl)phenyl]-3H-1-benzazepine-4-carboxamide, 2-Amino-N,N-dipropyl-8-[4-(pyrrolidine-1-carbonyl)phenyl]-3H-benzo[b]azepine-4-carboxamide, VTX 2337, VTX2337, VTX378

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H34N4O2
CAS Number:
Molecular Weight:
458.60
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

N4(CCCC4)C(=O)c1ccc(cc1)c2cc3c(cc2)C=C(C=C(N3)N)C(=O)N(CCC)CCC

InChI

1S/C28H34N4O2/c1-3-13-31(14-4-2)28(34)24-17-23-12-11-22(18-25(23)30-26(29)19-24)20-7-9-21(10-8-20)27(33)32-15-5-6-16-32/h7-12,17-19,30H,3-6,13-16,29H2,1-2H3

InChI key

CQAGPRVMJDWWEA-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective toll-like receptor 8 (TLR8) agonist that augments the lytic function of NK cells and enhances antibody-dependent cellular cytotoxicity.
Motolimod (VTX-2337) is a potent and selective human toll-like receptor 8 (TLR8) agonist (EC50 = 100 nM; no murine TLR8-stimulating potenty) with greatly reduced TLR7 potency (EC50 >30 µM) and little activity toward TLR3/4/5/8. Motolimod induces proinflammatory mediators production in huaman PBMCs (TNFα/IL12 EC50 = 140/120 nM) by stimulating TLR8-expressing monocytes and mDCs, but not non-TLR8-expressing pDCs. Motolimod augments the lytic function of NK cells and enhances antibody-dependent cellular cytotoxicity (ADCC).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library