Skip to Content
MilliporeSigma

SML4096

B022

≥97% (HPLC)

Synonym(s):

α-[2-[1-(2-Amino-5-chloro-4-pyrimidinyl)-2,3-dihydro-1H-indol-6-yl]ethynyl]-α-methyl-2-thiazolemethanol, 4-(1-(2-Amino-5-chloropyrimidin-4-yl)indolin-6-yl)-2-(thiazol-2-yl)but-3-yn-2-ol, NIK inhibitor Cmp1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H16ClN5OS
CAS Number:
Molecular Weight:
397.88
MDL number:
NACRES:
NA.21
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

ClC1=CN=C(N)N=C1N2C3=CC(C#CC(C4=NC=CS4)(C)O)=CC=C3CC2

Biochem/physiol Actions

Potent and selective NF-κB-inducing kinase (NIK) inhibitor that selectively disrupts NF-κB activation in cultures and in vivo.

B022 is an ATP site-targeting, potent and selective NF-κB-inducing kinase (NIK) inhibitor (Ki = 5 nM against 5 μM ATP using mouse NIK) that inhibits NIK-induced p100-to-p52 processing (IC50 <0.5 μM; Hepa1 cells overexpressing p100 & NIK) and NF-κB2 activity (by 38% by 0.5 μM and 96% by 5 μM B022; HEK293 NIK transfectant with NF-κB reporter plasmid). B022 protects mice from death due to liver-specific NIK overexpression in mice in vivo (30 mg/kg via b.i.d.tail vein injection).


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library