Skip to Content
MilliporeSigma

SML4103

4-Hydroperoxycyclophosphamide

≥98% (HPLC)

Synonym(s):

2-[Bis(2-chloroethyl)amino]tetrahydro-2-oxido-2H-1,3,2-oxazaphosphorin-4-yl hydroperoxide, 4-Hydroperoxycyclofosfamide, 4-OOH-CY, Asta 6496, Perfosfamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O4P
CAS Number:
Molecular Weight:
293.08
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Active metabolite of the nitrogen mustard cyclophosphamide.
4-Hydroperoxycyclophosphamide is the active metabolite of the nitrogen mustard cyclophosphamide. 4-Hydroperoxycyclophosphamide exhibits potent antineoplastic and immunosuppressive activities.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 1A - Skin Corr. 1B - Skin Sens. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library