Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C6H10NO3·K
CAS Number:
Molecular Weight:
183.25
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Quality Segment
assay
≥95% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
N-Hydroxypipecolic acid (1-Hydroxy-2-piperidinecarboxylic acid) is a plant metabolite that plays a crucial role in the establishment of systemic acquired resistance (SAR) in plants. It works in tandem with the immune signal salicylic acid to orchestrate SAR establishment. In response to pathogen attack, N-Hydroxypipecolic acid (NHP) accumulates systemically in the plant foliage and induces SAR (systemic acquired resistance) to bacterial and oomycete infection. NHP (N-Hydroxypipecolic acid) induces de novo systemic accumulation of extracellular nicotinamide adenine dinucleotide (phosphate) [eNAD(P)] mainly through the production of reactive oxygen species (ROS) by respiratory burst oxidase homolog F (RBOHF).
Disclaimer
Hygroscopic Store with desiccant
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
