Skip to Content
MilliporeSigma

SML4129

YM155

≥97% (HPLC)

Synonym(s):

1-(2-Methoxyethyl)-2-methyl-4,9-dioxo-3-(pyrazin-2-ylmethyl)-4,9-dihydro-1H-naphtho[2,3-d]imidazolium bromide, 11-(2-Methoxyethyl)-2-methyl-4,9-dioxo-3-(pyrazin-2-ylmethyl)-4,9-dihydro-1H-naphtho(2,3-d)imidazolium bromide, 4,9-Dihydro-1-(2-methoxyethyl)-2-methyl-4,9-dioxo-3-(pyrazinylmethyl)-1H-naphth[2,3-d]imidazolium bromide, Sepantronium bromide, Survivin Inhibitor I, Interleukin Enhancer-Binding Factor 3 Inhibitor, YM 155, YM-155

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H19N4O3·Br
CAS Number:
Molecular Weight:
443.29
MDL number:
NACRES:
NA.28
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Survivin expression inhibitor with anti-cancer efficacy in cultures and in vivo.
YM155 is a survivin expression inhibitor (effective conc. 10-100 nM for 6-48h; PC-3 & PCC-1 cells) that induces apoptosis in malignant melanoma and hormone-refractory prostate cancer (HRPC) cultures (GI50 = 2.3-11 nM) by targeting survivin gene promoter activity without affecting the expression of other IAPs or Bcl-2-related proteins even at concentrations as high as 100 nM. In a s.c. xenografted PC-3 tumor model in mice, 3-day continuous infusions of YM155 at 3 to 10 mg/kg induced massive tumor regression in vivo.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library