Skip to Content
MilliporeSigma

SML4138

VRK-IN-1

≥98% (HPLC)

Synonym(s):

4,4′-(2-Methylpyridine-3,5-diyl)bis(2,6-difluorophenol)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H11F4NO2
CAS Number:
Molecular Weight:
349.28
MDL number:
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC(C2=CN=C(C)C(C3=CC(F)=C(O)C(F)=C3)=C2)=CC(F)=C1O

Biochem/physiol Actions

Potent and selective vaccinia-related kinases 1 (VRK1) inhibitor that blocks VRK1-mediated histone H3 & H4 posttranslational epigenetic modifications.

VRK-IN-1 is a potent and selective vaccinia-related kinases 1 (VRK1) inhibitor (VRK1 IC50 = 150 nM; % inhibition at 1 µM = 78/MAP4K4, 53/CAMKK2, and <25 against 46 other kinases). VRK1 is reported to modulate histone H3 & H4 posttranslational epigenetic modifications, VRK-IN-1 treatment downregulates H4K16ac (by 36%, 45%, 73%, respectively, post 24h incubation with 200, 400, 600 nM VRK-IN-1 in serum-deprived A549 cultures) and H3K4me3 (by 70% post 24h 600 nM treatment; A549 cells), while upregulates H3K9me3 (by 200% post 24h 600 nM treatment; A549 cells).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library