Skip to Content
MilliporeSigma

SML4141

KCNQ2 Activator Ebio1

≥98% (HPLC)

Synonym(s):

Ebio1, N-(1,2-Dihydro-5-acenaphthylenyl)-4-fluorobenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H14FNO
CAS Number:
Molecular Weight:
291.32
MDL number:
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(C(NC2=CC=C3C4=C2C=CC=C4CC3)=O)C=C1

Biochem/physiol Actions

Uniquely acting, highly selective sub-type activator for the KCNQ2 (KV7.2) potassium channel.
Ebio1 is a potent, sub-type selective KCNQ2 (KV7.2) potassium channel direct activator that is distinct from previously known activators of voltage-gated ion channels. Ebio1 operates through a unique mechanism, inducing a twist-to-open movement in the S6 helices of the channel, thereby increasing both the open probability and conductance of KCNQ2. KCNQ2 Activator Ebio1 has an EC50 of 247.3 nM for KCNQ2, making it tenfold more selective compared to KCNQ2/3, KCNQ4, and KCNQ5. Moreover, Ebio1 demonstrates minimal to negligible activation effects on other tested Kv, Nav, Cav, and K2P channel members, including hERG, BK, Nav1.1, Cav2.1, and TREK1.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library