Skip to Content
MilliporeSigma

SML4146

Galidesivir Hydrochloride

≥95% (HPLC)

Synonym(s):

(2S,3S,4R,5R)-2-(4-Amino-5H-pyrrolo[3,2-d]pyrimidin-7-yl)-5-(hydroxymethyl)pyrrolidine-3,4-diol hydrochloride, Immucillin A hydrochloride, BCX4430 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15N5O3 · xHCl
Molecular Weight:
265.27 (free base basis)
UNSPSC Code:
12352203
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Galidesivir (BCX4430) is an inhibitor of viral RNA-dependent RNA polymerase (RDRP) function with broad spectrum antiviral activity including against West Nile virus and tick-borne flaviviruses, Zika, haemorrhagic filoviruses Ebola and Marburg in addition to bunyaviruses, arenaviruses, paramyxoviruses, and coronaviruses. Galidesivir potently inhibits SARS-CoV-2 replication in cell-based assays. Also, it protected against Ebola virus and Marburg virus disease in rodent models and protected macaques from Marburg virus infection when given as late as 48 hours after infection.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library