Skip to Content
MilliporeSigma

SML4147

Cyclopropavir

≥98% (HPLC), powder

Synonym(s):

2-Amino-9-[(Z)-[2,2-bis(hydroxymethyl)cyclopropylidene]methyl]-1,9-dihydro-6H-purin-6-one, (Z)-9-((2,2-bis-(hydroxymethyl)cyclopropylidene)methyl)guanine, Filociclovir, ZSM-I-62, MBX-400

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H13N5O3
CAS Number:
Molecular Weight:
263.25
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cyclopropavir (Filociclovir) is an orally available and potent antiviral agent that is effective against human and murine cytomegaloviruses (MCMV and HCMV) infection. It has an in vitro median effective concentration (EC50) of approximately 1.2 μM for wild-type CMV strains. Notably, for ganciclovir-resistant strains, cyclopropavir EC50 values are typically lower than those of ganciclovir, ranging from 0.04 to 37.2μM, often demonstrating a 5 to 10-fold or greater advantage. Cyclopropavir, a methylenecyclopropane nucleoside analog , upon activation by phosphorylation targets HCMV DNA polymerase and terminates viral DNA synthesis. It is also effective against human herpes simplex virus 6 (HSV-6) and human herpes simplex virus 8 (HSV-8).


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library