Skip to Content
MilliporeSigma

SML4150

Ciprofloxacin hydrochloride hydrate

≥98% (HPLC), powder

Synonym(s):

1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride hydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid monohydrochloride, monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H18FN3O3·HCl · xH2O
CAS Number:
Molecular Weight:
367.80 (anhydrous basis)
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(C1=CN(C2CC2)C3=CC(N4CCNCC4)=C(C=C3C1=O)F)O.Cl.[xH2O]

Application

Ciprofloxacin hydrochloride hydrate is suitable for use in research to study antibiotic resistance mechanisms. It is also used to inhibit cell proliferation and to sensitize cells to TRAIL (TNF-related apoptosis-inducing ligand) -Induced apotosis.

Biochem/physiol Actions

Ciprofloxacinis a fluoroquinolone antibiotic with broad-spectrum activity, which killsbacteria by blocking DNA gyrase and type IV topoisomerase—critical enzymes forDNA replication and transcription—leading to cell death. It is effectiveagainst a broad array of both Gram-positive and Gram-negative bacteria, inaddition to Mycoplasmas. While the hydrochloride form of ciprofloxacin ishighly soluble in water, ciprofloxacin itself is only sparingly soluble inaqueous solutions.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library