Skip to Content
MilliporeSigma

SML4152

Ipragliflozin

≥98% (HPLC)

Synonym(s):

(1S)-1,5-Anhydro-1-C-[3-(benzo[b]thien-2-ylmethyl)-4-fluorophenyl]-D-glucitol, (2S,3R,4R,5S,6R)-2-(3-(Benzo[b]thien-2-ylmethyl)-4-fluorophenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, ASP-1941

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H21FO5S
CAS Number:
Molecular Weight:
404.45
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@H]1[C@@H](O[C@H](CO)[C@@H](O)[C@@H]1O)C2=CC(CC3=CC4=CC=CC=C4S3)=C(F)C=C2

Biochem/physiol Actions

Ipragliflozin is a potent and highly selective sodium-glucose cotransporter-2 (SGLT2) inhibitor, demonstrating excellent oral bioavailability with an IC50 of 1,876 nM for SGLT1and 7.38 nM for hSGLT2. Ipragliflozin features dose-dependent pharmacokinetics, effectively induces glucosuria, and has been shown to decrease blood glucose levels, reduce blood pressure, lower body weight, and diminish both hypoglycaemic risk and abdominal symptoms in diabetic patients. Additionally, Ipragliflozin increases insulin levels in the pancreas and prevents the loss of insulin-positive cells in islets of db/db mice. ASP-1941 also maintains a favourable safety profile as monotherapy and is compatible with other hypoglycaemic agents, such as sulphonyl urea, metformin, pioglitazone, and dipeptidyl peptidase-4 (DPP4) inhibitors. Ipragliflozin has been considered as an anti-obesity and anti-diabetic agent.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library