Skip to Content
MilliporeSigma

SML4154

Pemafibrate

≥98% (HPLC), powder

Synonym(s):

(2R)-2-[3-[[2-Benzoxazolyl[3-(4-methoxyphenoxy)propyl]amino]methyl]phenoxy]butanoic acid, (R)-2-[3-[[Benzoxazol-2-yl[3-(4-methoxyphenoxy)propyl]amino]methyl]phenoxy]butanoic acid, K 13675, K 877, K-13675, K-877, K13675, K877, (R)-K13675, (R)-K-13675, (R)-K13675

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H30N2O6
CAS Number:
Molecular Weight:
490.55
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

COC(C=C1)=CC=C1OCCCN(CC2=CC(O[C@H](CC)C(O)=O)=CC=C2)C3=NC4=C(O3)C=CC=C4

Application

Pemafibrate may be used in research studies tostudy its effects on lipid metabolism, inflammation, and cardiovascular health.

Biochem/physiol Actions

Pemafibrate is an orally active, highly potent and selective peroxisome proliferator-activated receptor alpha (PPARα) agonist (hPPAR EC50 = 1 nM/α, 1.10 μM/γ, 1,58 μM/δ by cell-based reporter assays using respective transfectants). When administered in vivo Pemafibrate is >100-fold more effective than fenofibrate in lowering triglyceride (TG) in normal rats (1 mg/kg p.o.) and in increasing HDL-cholesterol (HDL-C) in human apoA-I (h-apoA-I) transgenic mice (3 mg/kg p.o.). Compared to other fibrates, pemafibrate′s distinct Y-shaped structure aids in stronger PPAR-α activation, better lipid binding, greater potency, and adaptability to structural changes caused by binding of coactivators. Pemafibrate shows anti-inflammatory activities like lowering the levels of interleukin-6, tumor necrosis factor-α, monocyte chemoattractant protein-1, vascular cell adhesion molecule-1 (VCAM-1), macrophage marker F4/80). It also exhibits antiatherogenic effects by lowering the levels of certain factors such as plasma triglycerides (TG), TG-rich lipoproteins, and apolipoprotein C-III (apoC-III), and increases lipoprotein lipase (LPL) activity and high-density lipoprotein (HDL) levels. Pemafibrate promotes fibroblast growth factor 21 and contributes to the reduction of insulin resistance.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library