Skip to Content
MilliporeSigma

SML4155

Detomidine

≥98% (HPLC)

Synonym(s):

4-(2,3-Dimethylbenzyl)imidazole, 4-((2,3-Dimethylphenyl)methyl)-1H-imidazole, 5-(2,3-Dimethylbenzyl)-1H-imidazole, 5-[(2,3-Dimethylphenyl)methyl]-1H-imidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14N2
CAS Number:
Molecular Weight:
186.25
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=C(C=CC=C1C)CC2=CN=CN2

Application

Detomidine may be used in the research field foranimal behavior studies to facilitate sedation and to conduct procedures thatneed immobilization of animals for observation.

Biochem/physiol Actions

Detomidine is an α2-adrenergic receptor(α2-adrenoceptor) agonist with in vivo sedative and analgesic efficacy.In receptor binding experiments, detomidine showed strong affinity to alpha2-receptors. There was some binding affinity towards alpha 1-receptors also butonly negligible or no affinity towards dopamine, opiate and adenosinereceptors. Detomidine is a derivative of imidazole that exerts muscle relaxation effects. It also acts as an anesthetic adjunct. Detomidine also shows cardiovascular effectssuch as induction of a state of hypertension, bradycardia and increasedsystemic and pulmonary vascular resistance.(4)


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library