Skip to Content
MilliporeSigma

SML4156

SiR-DBCO

≥90% (HPLC)

Synonym(s):

[10-[5-[[3-(2-azatricyclo[10.4.0.04,9]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl)-3-oxopropyl]carbamoyl]-2-carboxyphenyl]-7-(dimethylamino)-5,5-dimethylbenzo[b][1]benzosilin-3-ylidene]-dimethylazanium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H42N4O4Si
CAS Number:
Molecular Weight:
730.92
UNSPSC Code:
12352200
Assay:
≥90% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

SiR-DBCO is a cell-permeable, silicon-rhodamine strained alkyne click reagent, essential in forming the MAO-SiR complex when used with MAO-N3 for mitochondrial imaging. In studies, this combination showed remarkable stability in HeLa cells, maintaining signal integrity over 12.5 hours with minimal decay, a clear indication of effective mitochondrial labeling. This contrasts sharply with traditional dyes like MitoTracker Deep Red, which often exhibit significant signal loss within hours. Thus, SiR-DBCO, in conjunction with MAO-N3, enhances the duration and quality of mitochondrial imaging, even under lower-intensity lighting conditions. Note: In time-lapse imaging, cells treated with 200 nM MAO-N3 and 100 nM SiR-DBCO, collectively known as MAO-SiR, exhibit minimal signal degradation and a rise in cell density, indicative of robust mitochondrial function and cell health. This outcome is in stark contrast to the results observed with dyes like MitoTracker Deep Red, which typically show a substantial decline in signal intensity within just a few hours.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library