Skip to Content
MilliporeSigma

SML4157

FINO2 10% solution in DMSO

≥98% (HPLC), solution

Synonym(s):

rel-(5α,8α)-8-(1,1-Dimethylethyl)-3-methyl-1,2-dioxaspiro[4.5]decane-3-ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H28O3
Molecular Weight:
256.38
MDL number:
UNSPSC Code:
12352203
Assay:
≥98% (HPLC)
Form:
solution
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solution

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

FINO2 is used as aresearch tool to study pathways of ferroptosis and to probe explore how thesepathways relate to diseases including Alzheimer’s, Parkinson’s andcardiovascular diseases.

Biochem/physiol Actions

FINO2 is a novel inducer of ferroptosis. It acts in a different manner than class 1 inducers, which deplete glutathione by inhibiting cystine import via system xc−, or class 2 inihbitors, which covalently inhibit glutathione peroxidase 4 (GPX4). FINO2 has a two-fold mechanism of action. It indirectly inhibits the enzymatic of glutathione peroxidase 4 (GPX4) and directly oxidizes iron, leading to widespread lipid peroxidation and eventual cell death. FINO2 was found to be more potent in cancer cells than in nonmalignant cells of the same type.


Pictograms

Flame

Signal Word

Danger

Hazard Codes

Hazard Classifications

Org. Perox. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library